(6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione 98%
Supplier: Ambeed
|
Warning
|
A215860-10MG
A215860-1G
A215860-250MG
A215860-25MG
A215860-50MG
A215860-5G
A215860-5MG
77152-616EA
6.18
USD
77152-616
77152-618
77152-620
77152-622
77152-624
77152-626
77152-628
(6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione 98%
(6R,12aR)-6-(Benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
Precaución: for Research Use Only
Formula:
C₂₁H₁₇N₃O₄ MW: 375.37 g/mol |
MDL Number:
MFCD19443817 CAS Number: 171596-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...