2-Chloronicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-cyanopyridine, 2-Chloropyridine-3-carbonitrile

A215332-100G A215332-10G A215332-25G A215332-500G
77152-592EA 15.45 USD
77152-592 77152-594 77152-596 77152-598
2-Chloronicotinonitrile 98%
2-Chloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
MDL Number: MFCD00014628
CAS Number: 6602-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR