5-tert-Butyl 2-ethyl 6,7-dihydrofuro[3,2-c]pyridine-2,5(4H)-dicarboxylate 95+%

Supplier: Ambeed
Warning

A215261-100MG A215261-1G A215261-250MG
77152-588EA 67 USD
77152-588 77152-590 77685-579
5-tert-Butyl 2-ethyl 6,7-dihydrofuro[3,2-c]pyridine-2,5(4H)-dicarboxylate 95+%
5-tert-Butyl 2-ethyl 6,7-dihydrofuro[3,2-c]pyridine-2,5(4H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MDL Number: MFCD10698557
CAS Number: 1060814-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR