Methyl 6-bromo-1-methyl-1H-indole-4-carboxylate 98+%

Supplier: Ambeed
Warning

A215126-1G A215126-250MG A215126-5G
77152-582EA 39 USD
77152-582 77152-584 77152-586
Methyl 6-bromo-1-methyl-1H-indole-4-carboxylate 98+%
Methyl 6-bromo-1-methyl-1H-indole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MDL Number: MFCD28641837
CAS Number: 1090903-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR