2-(2-Methoxyphenoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A214656-100MG A214656-1G A214656-250MG
77152-566EA 54.99 USD
77152-566 77152-568 77152-570
2-(2-Methoxyphenoxy)benzoic acid 97%
2-(2-Methoxyphenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MDL Number: MFCD02220804
CAS Number: 21905-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR