UNC2881 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(4-(1H-Imidazol-1-yl)benzyl)-2-(butylamino)-4-((trans-4-hydroxycyclohexyl)amino)pyrimidine-5-carboxamide, AOB6195, N-(4-(1H-imidazol-1-yl)benzyl)-2-(butylamino)-4-(((1r,4r)-4-hydroxycyclohexyl)amino)pyrimidine-5-carboxamide
A214490-10MG
A214490-1MG
A214490-5MG
77152-554EA
153
USD
77152-554
77152-556
77152-558
UNC2881 98+%
UNC2881
Precaución: for Research Use Only
Formula:
C₂₅H₃₃N₇O₂ MW: 463.58 g/mol |
MDL Number:
MFCD28142814 CAS Number: 1493764-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...