5-(3,4-Dichlorophenyl)-1,3,4-thiadiazol-2-amine 95%
Supplier: Ambeed
|
Warning
|
A213958-10G
A213958-1G
77152-534EA
226
USD
77152-534
77900-621
5-(3,4-Dichlorophenyl)-1,3,4-thiadiazol-2-amine 95%
5-(3,4-Dichlorophenyl)-1,3,4-thiadiazol-2-amine
Precaución: for Research Use Only
Formula:
C₈H₅Cl₂N₃S |
MDL Number:
MFCD01191258 CAS Number: 28004-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...