2,4-Dibromo-5-fluoropyrimidine 95%

Supplier: Ambeed
Warning

A213853-100G A213853-1G A213853-250MG A213853-25G A213853-5G
77152-524EA 386.25 USD
77152-524 77152-526 77152-528 77152-530 77152-532
2,4-Dibromo-5-fluoropyrimidine 95%
2,4-Dibromo-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄HBr₂FN₂
MW: 255.871 g/mol
MDL Number: MFCD21647860
CAS Number: 1372096-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR