Dorsmorphin dihydrochloride 99+%

Supplier: Ambeed
Warning

A213616-100MG A213616-1MG A213616-250MG A213616-50MG A213616-5MG A213616-10MG
77152-514EA 219 USD
77152-514 77152-516 77152-518 77152-520 77152-522 77612-104
Dorsmorphin dihydrochloride 99+%
Dorsmorphin dihydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₇Cl₂N₅O
MDL Number: MFCD11112197
CAS Number: 1219168-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR