1-Butyl-4-methylpyridinium chloride 98%

Supplier: Ambeed
Warning

A213208-100G A213208-1G A213208-25G A213208-5G
77152-484EA 189 USD
77152-484 77152-486 77152-488 77152-490
1-Butyl-4-methylpyridinium chloride 98%
1-Butyl-4-methylpyridinium chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClN
MDL Number: MFCD03095439
CAS Number: 112400-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR