3-Bromo-2-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-hydroxynaphthalene, 3-Bromonaphthalen-2-ol

A212214-1G A212214-250MG A212214-5G
77152-464EA 50.47 USD
77152-464 77152-466 77152-468
3-Bromo-2-naphthol 98%
3-Bromo-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
MDL Number: MFCD10000951
CAS Number: 30478-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR