2-Hydroxyacetophenone 97%
Supplier: Ambeed
|
Warning
|
A212172-100G
A212172-10G
A212172-25G
A212172-500G
A212172-5G
77152-454EA
118
USD
77152-454
77152-456
77152-458
77152-460
77152-462
2-Hydroxyacetophenone 97%
2-Hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₈O₂ |
MDL Number:
MFCD00041829 CAS Number: 582-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...