UNC0379 98%

Supplier: Ambeed
Warning

Synonyms: 6,7-Dimethoxy-2-(pyrrolidin-1-yl)-N-(5-(pyrrolidin-1-yl)pentyl)quinazolin-4-amine

A211708-10MG A211708-1MG A211708-5MG A211708-100MG A211708-50MG
77152-448EA 80 USD
77152-448 77152-450 77152-452 77901-832 77901-859
UNC0379 98%
UNC0379

Precaución: for Research Use Only

Formula: C₂₃H₃₅N₅O₂
MW: 413.56 g/mol
MDL Number: MFCD28167817
CAS Number: 1620401-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR