(S)-2-(1-(2-(2,3-Dihydrobenzofuran-5-yl)ethyl)pyrrolidin-3-yl)-2,2-diphenylacetamide hydrobromide 98%
Supplier: Ambeed
|
Warning
|
A211196-1G
A211196-250MG
A211196-25MG
A211196-50MG
A211196-10MG
A211196-5G
A211196-100MG
A211196-25G
77152-440EA
54
USD
77152-440
77152-442
77152-444
77152-446
77685-553
77685-554
77900-654
77901-155
(S)-2-(1-(2-(2,3-Dihydrobenzofuran-5-yl)ethyl)pyrrolidin-3-yl)-2,2-diphenylacetamide hydrobromide 98%
(S)-2-(1-(2-(2,3-Dihydrobenzofuran-5-yl)ethyl)pyrrolidin-3-yl)-2,2-diphenylacetamide hydrobromide
Precaución: for Research Use Only
Formula:
C₂₈H₃₁BrN₂O₂ |
MDL Number:
MFCD08141803 CAS Number: 133099-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...