5,6-Dibromo-1,2-dihydroacenaphthylene 95%

Supplier: Ambeed
Warning

A210921-10G A210921-1G A210921-250MG A210921-5G
77152-432EA 253 USD
77152-432 77152-434 77152-436 77152-438
5,6-Dibromo-1,2-dihydroacenaphthylene 95%
5,6-Dibromo-1,2-dihydroacenaphthylene

Precaución: for Research Use Only

Formula: C₁₂H₈Br₂
MDL Number: MFCD00191989
CAS Number: 19190-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR