Cyclopropyl-2-methylphenyl ketone 95%

Supplier: Ambeed
Warning

Synonyms: Cyclopropyl 2-methylphenyl ketone

A210702-100MG A210702-1G A210702-250MG
77152-426EA 37.08 USD
77152-426 77152-428 77152-430
Cyclopropyl-2-methylphenyl ketone 95%
Cyclopropyl-2-methylphenyl ketone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
MDL Number: MFCD03841206
CAS Number: 39615-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR