Bis(3-ethynylbenzyl)amine 97%

Supplier: Ambeed
Warning

A210127-100MG A210127-250MG
77152-408EA 14.42 USD
77152-408 77152-410
Bis(3-ethynylbenzyl)amine 97%
Bis(3-ethynylbenzyl)amine

Precaución: for Research Use Only

Formula: C₁₈H₁₅N
MW: 245.31 g/mol
MDL Number: MFCD29044845
CAS Number: 1825394-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR