6-(2-Chloro-5-fluoropyrimidin-4-yl)-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole 97%
Supplier: Ambeed
|
Warning
|
A209941-100MG
A209941-10G
A209941-1G
A209941-250MG
A209941-5G
A209941-25G
77152-396EA
5
USD
77152-396
77152-398
77152-400
77152-402
77152-404
77892-315
6-(2-Chloro-5-fluoropyrimidin-4-yl)-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole 97%
6-(2-Chloro-5-fluoropyrimidin-4-yl)-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole
Precaución: for Research Use Only
Formula:
C₁₅H₁₃ClF₂N₄ |
MDL Number:
MFCD25977328 CAS Number: 1231930-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...