2-Chloro-3-iodo-4-methoxypyridine 95%

Supplier: Ambeed
Warning

A208428-100MG A208428-10G A208428-1G A208428-250MG A208428-25G A208428-5G
77152-374EA 16 USD
77152-374 77152-376 77152-378 77152-380 77152-382 77152-384
2-Chloro-3-iodo-4-methoxypyridine 95%
2-Chloro-3-iodo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
MDL Number: MFCD18415710
CAS Number: 1163693-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR