Phenethyltrimethoxysilane 95%

Supplier: Ambeed
Warning

Synonyms: Trimethoxy(phenethyl)silane, Trimethoxy(2-phenylethyl)silane [contains ca. 25% Trimethoxy(1-phenylethyl)silane], Trimethoxy(2-phenylethyl)silane

A206846-100G A206846-25G A206846-5G
77152-338EA 200.85 USD
77152-338 77152-340 77152-342
Phenethyltrimethoxysilane 95%
Phenethyltrimethoxysilane

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₃Si
MW: 226.347 g/mol
MDL Number: MFCD00053852
CAS Number: 49539-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR