6-Mercapto-6-deoxy-β-Cyclodextrin 95%

Supplier: Ambeed
Warning

A206653-100MG A206653-1G A206653-250MG A206653-5G
77152-324EA 75.19 USD
77152-324 77152-326 77152-328 77685-533
6-Mercapto-6-deoxy-β-Cyclodextrin 95%
6-Mercapto-6-deoxy-β-Cyclodextrin

Precaución: for Research Use Only

Formula: C₄₂H₇₀O₃₄S
MW: 1151.04 g/mol
MDL Number: MFCD31537208
CAS Number: 81644-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR