Amuvatinib 98%

Supplier: Ambeed
Warning

Synonyms: N-(Benzo[d][1,3]dioxol-5-ylmethyl)-4-(benzofuro[3,2-d]pyrimidin-4-yl)piperazine-1-carbothioamide, MP-470

A205668-10MG A205668-1MG A205668-50MG A205668-5MG
77152-306EA 67.98 USD
77152-306 77152-308 77152-310 77152-312
Amuvatinib 98%
Amuvatinib

Precaución: for Research Use Only

Formula: C₂₃H₂₁N₅O₃S
MW: 447.5 g/mol
MDL Number: MFCD16038298
CAS Number: 850879-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR