7-(tert-Butoxycarbonyl)- 5,6,7,8-tetrahydroimidazo-[1,5-a]pyrazine-1-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A204655-100MG
A204655-250MG
77152-284EA
226.6
USD
77152-284
77152-286
7-(tert-Butoxycarbonyl)- 5,6,7,8-tetrahydroimidazo-[1,5-a]pyrazine-1-carboxylic acid 98%
7-(tert-Butoxycarbonyl)- 5,6,7,8-tetrahydroimidazo-[1,5-a]pyrazine-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₇N₃O₄ MW: 267.28 g/mol |
MDL Number:
MFCD12198789 CAS Number: 1160248-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...