Methyl 2-bromo-5-fluoroisonicotinate 98%

Supplier: Ambeed
Warning

A204547-100MG A204547-1G A204547-250MG A204547-5G A204547-25G
77152-278EA 5 USD
77152-278 77152-280 77152-282 77567-962 77901-940
Methyl 2-bromo-5-fluoroisonicotinate 98%
Methyl 2-bromo-5-fluoroisonicotinate

Precaución: For research use only.

Formula: C₇H₅BrFNO₂
MDL Number: MFCD09951945
CAS Number: 885588-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR