Methyl 4-amino-3-(trifluoromethoxy)benzoate 95%

Supplier: Ambeed
Warning

A202043-10G A202043-1G A202043-250MG A202043-25G A202043-5G
77152-208EA 130 USD
77152-208 77152-210 77152-212 77152-214 77685-511
Methyl 4-amino-3-(trifluoromethoxy)benzoate 95%
Methyl 4-amino-3-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₃
MDL Number: MFCD15527567
CAS Number: 457097-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR