1-Bromo-2,5-difluoro-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Brom-2,5-difluor-4-nitrobenzol, 4-Bromo-2,5-difluoronitrobenzene

A202018-100G A202018-10G A202018-1G A202018-25G A202018-5G
77152-198EA 168.92 USD
77152-198 77152-200 77152-202 77152-204 77152-206
1-Bromo-2,5-difluoro-4-nitrobenzene 98%
1-Bromo-2,5-difluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
MDL Number: MFCD00042184
CAS Number: 167415-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR