(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one (Mixture of A and B) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Aloin (Mixture of A and B), (S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one(Mixture of A and B)
A201696-100G
A201696-1G
A201696-25G
A201696-5G
77152-188EA
337.84
USD
77152-188
77152-190
77152-192
77152-194
(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one (Mixture of A and B) 95%
(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one (Mixture of A and B)
Precaución: for Research Use Only
Formula:
C₂₁H₂₂O₉ MW: 418.398 g/mol |
MDL Number:
MFCD00151160 CAS Number: 8015-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...