3-Fluoro-4-(trifluoromethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A201341-100MG A201341-1G A201341-250MG
77152-182EA 26.78 USD
77152-182 77152-184 77152-186
3-Fluoro-4-(trifluoromethoxy)benzonitrile 97%
3-Fluoro-4-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₄NO
MW: 205.1091 g/mol
MDL Number: MFCD06660203
CAS Number: 886498-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR