D-Threonine benzyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3S)-Benzyl 2-amino-3-hydroxybutanoate hydrochloride, H-D-Thr-OBzl.HCl, D-Threonine Benzyl Ester HCl

A201294-100MG A201294-1G A201294-250MG A201294-25G A201294-5G
77152-164EA 5.15 USD
77152-164 77152-166 77152-168 77152-170 77152-172
D-Threonine benzyl ester hydrochloride 97%
D-Threonine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 245.70 g/mol
MDL Number: MFCD28975347
CAS Number: 75748-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR