(S)-Pyrrolidine-3-carbonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A201195-100MG A201195-250MG A201195-1G
77152-160EA 64.89 USD
77152-160 77152-162 77611-952
(S)-Pyrrolidine-3-carbonitrile hydrochloride 95%
(S)-Pyrrolidine-3-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂
MDL Number: MFCD11656840
CAS Number: 1153950-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR