4-(3,4-Dichlorobenzyl)piperidine hydrochloride 98%

Supplier: Ambeed
Warning

A200829-100MG A200829-1G A200829-250MG A200829-5G
77152-138EA 20 USD
77152-138 77152-140 77152-142 77152-144
4-(3,4-Dichlorobenzyl)piperidine hydrochloride 98%
4-(3,4-Dichlorobenzyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂N·ClH
MDL Number: MFCD03840139
CAS Number: 1171138-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR