(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A199968-100MG A199968-1G A199968-250MG A199968-25G A199968-5G
77152-116EA 6.18 USD
77152-116 77152-118 77152-120 77152-122 77152-124
(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 95%
(R)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂N·ClH
MW: 219.659 g/mol
MDL Number: MFCD08751413
CAS Number: 1218935-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR