1-Butyl-3-methylimidazolium iodide 96%

Supplier: Ambeed
Warning

A199824-100G A199824-10G A199824-25G A199824-5G
77152-108EA 152.44 USD
77152-108 77152-110 77152-112 77152-114
1-Butyl-3-methylimidazolium iodide 96%
1-Butyl-3-methylimidazolium iodide

Precaución: for Research Use Only

Formula: C₈H₁₅IN₂
MW: 266.12 g/mol
MDL Number: MFCD08276307
CAS Number: 65039-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR