Pyridin-4-ylmethanol hydrobromide 97%

Supplier: Ambeed
Warning

A199249-25G A199249-5G
77152-092EA 40 USD
77152-092 77152-094
Pyridin-4-ylmethanol hydrobromide 97%
Pyridin-4-ylmethanol hydrobromide

Precaución: for Research Use Only

Formula: C₆H₈BrNO
MW: 190.04 g/mol
MDL Number: MFCD29047074
CAS Number: 65737-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR