3-(Trifluoroacetyl)indole 98%

Supplier: Ambeed
Warning

A198789-100MG A198789-1G A198789-250MG A198789-5G
77152-078EA 7.21 USD
77152-078 77152-080 77152-082 77152-084
3-(Trifluoroacetyl)indole 98%
3-(Trifluoroacetyl)indole

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
MDL Number: MFCD00182114
CAS Number: 14618-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR