9-Bromo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine 98%

Supplier: Ambeed
Warning

A198692-250MG A198692-1G
77152-076EA 40 USD
77152-076 77593-288
9-Bromo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine 98%
9-Bromo-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O
MW: 265.10 g/mol
MDL Number: MFCD22372760
CAS Number: 1282516-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR