2,5-dichloro-N-(5-methyl-1H-pyrazol-3-yl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A198401-100MG A198401-250MG
77152-072EA 13.39 USD
77152-072 77152-074
2,5-dichloro-N-(5-methyl-1H-pyrazol-3-yl)pyrimidin-4-amine 97%
2,5-dichloro-N-(5-methyl-1H-pyrazol-3-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₈H₇Cl₂N₅
MW: 244.08 g/mol
MDL Number: MFCD12406889
CAS Number: 543712-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR