10H-Spiro[acridine-9,9'-fluorene] 98%

Supplier: Ambeed
Warning

A197840-100MG A197840-1G A197840-250MG
77152-066EA 21 USD
77152-066 77152-068 77152-070
10H-Spiro[acridine-9,9'-fluorene] 98%
10H-Spiro[acridine-9,9'-fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₇N
MW: 331.42 g/mol
MDL Number: MFCD29919320
CAS Number: 92638-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR