NMS-873 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(Cyclopentylthio)-5-(((2-methyl-4'-(methylsulphonyl)-[1,1'-biphenyl]-4-yl)oxy)methyl)-4H-1,2,4-triazol-4-yl)pyridine

A197765-100MG A197765-10MG A197765-1MG A197765-50MG A197765-5MG
77152-056EA 709.67 USD
77152-056 77152-058 77152-060 77152-062 77152-064
NMS-873 98%
NMS-873

Precaución: for Research Use Only

Formula: C₂₇H₂₈N₄O₃S₂
MW: 520.67 g/mol
MDL Number: MFCD28009371
CAS Number: 1418013-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR