L(+)-Leucine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
A197119-1000G
A197119-100G
A197119-25G
A197119-500G
77152-038EA
51
USD
77152-038
77152-040
77152-042
77152-044
L(+)-Leucine 97%
L(+)-Leucine
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Melting Pt: 300 |
MDL Number:
MFCD00002617 CAS Number: 61-90-5 Merck Index: 14,05451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...