3-(5,6-Dichloro-1H-benzo[d]imidazol-2-yl)propanol 97%

Supplier: Ambeed
Warning

A196739-100MG A196739-1G A196739-250MG
77152-032EA 54.59 USD
77152-032 77152-034 77152-036
3-(5,6-Dichloro-1H-benzo[d]imidazol-2-yl)propanol 97%
3-(5,6-Dichloro-1H-benzo[d]imidazol-2-yl)propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₂O
MW: 245.10 g/mol
MDL Number: MFCD11106864
CAS Number: 6478-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR