5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A196116-10G A196116-1G A196116-250MG A196116-5G
77152-016EA 288 USD
77152-016 77152-018 77152-020 77152-022
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD27987377
CAS Number: 1638761-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR