Abexinostat 98%

Supplier: Ambeed
Warning

A195616-10MG A195616-1MG A195616-25MG A195616-50MG A195616-5MG A195616-100MG A195616-1G A195616-250MG
77151-998EA 39 USD
77151-998 77152-000 77152-002 77152-004 77152-006 77611-880 77685-484 77685-485
Abexinostat 98%
Abexinostat

Precaución: for Research Use Only

Formula: C₂₁H₂₃N₃O₅
MDL Number: MFCD10565969
CAS Number: 783355-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR