7-Bromo-5-chloro-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A195115-1G A195115-250MG A195115-5G
77151-986EA 21 USD
77151-986 77151-988 77892-261
7-Bromo-5-chloro-1H-indole-2-carboxylic acid 97%
7-Bromo-5-chloro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅BrClNO₂
MDL Number: MFCD08752934
CAS Number: 952959-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR