(R)-Cyclobutyl(phenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A193872-100MG A193872-250MG
77151-968EA 78.28 USD
77151-968 77151-970
(R)-Cyclobutyl(phenyl)methanamine hydrochloride 97%
(R)-Cyclobutyl(phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClN
MW: 197.7 g/mol
MDL Number: MFCD24419176
CAS Number: 1956435-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR