(R)-Methyl 2-amino-3-methoxypropanoate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-OMe-Ser-OMe·HCl
A193799-100MG
A193799-10G
A193799-1G
A193799-250MG
A193799-5G
A193799-100G
A193799-25G
77151-958EA
5
USD
77151-958
77151-960
77151-962
77151-964
77151-966
77892-252
77892-253
(R)-Methyl 2-amino-3-methoxypropanoate hydrochloride 95%
(R)-Methyl 2-amino-3-methoxypropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₃ |
MDL Number:
MFCD29044850 CAS Number: 1800300-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...