2-Fluoro-N-methyl-4-(7-(quinolin-6-ylmethyl)imidazo[1,2-b][1,2,4]triazin-2-yl)benzamide 99%

Supplier: Ambeed
Warning

Synonyms: Capmatinib

A193720-1MG A193720-5MG A193720-100MG A193720-250MG A193720-1G
77151-954EA 14 USD
77151-954 77151-956 77600-568 77600-570 77685-470
2-Fluoro-N-methyl-4-(7-(quinolin-6-ylmethyl)imidazo[1,2-b][1,2,4]triazin-2-yl)benzamide 99%
2-Fluoro-N-methyl-4-(7-(quinolin-6-ylmethyl)imidazo[1,2-b][1,2,4]triazin-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₃H₁₇FN₆O
MW: 412.42 g/mol
MDL Number: MFCD18633285
CAS Number: 1029712-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR