8-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine 95%

Supplier: Ambeed
Warning

A193383-1G A193383-250MG A193383-5G A193383-100MG
77151-948EA 80 USD
77151-948 77151-950 77151-952 77892-250
8-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine 95%
8-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD15144595
CAS Number: 54230-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR