(S)-7-Hydroxy-2-(4-hydroxyphenyl)-6-(3-methylbut-2-en-1-yl)chroman-4-one 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bavachin
A193314-1MG
A193314-5MG
77151-940EA
11.33
USD
77151-940
77151-942
(S)-7-Hydroxy-2-(4-hydroxyphenyl)-6-(3-methylbut-2-en-1-yl)chroman-4-one 99+%
(S)-7-Hydroxy-2-(4-hydroxyphenyl)-6-(3-methylbut-2-en-1-yl)chroman-4-one
Precaución: for Research Use Only
Formula:
C₂₀H₂₀O₄ MW: 324.37 g/mol |
MDL Number:
MFCD11617359 CAS Number: 19879-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...