2-Chloro-5-fluoro-4-(methylsulfanyl)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoro-4-(methylthio)pyrimidine

A192678-1G A192678-5G A192678-250MG
77151-932EA 22 USD
77151-932 77151-934 77892-244
2-Chloro-5-fluoro-4-(methylsulfanyl)pyrimidine 97%
2-Chloro-5-fluoro-4-(methylsulfanyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂S
MW: 178.615 g/mol
MDL Number: MFCD16876993
CAS Number: 87789-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR